siRNA Details
| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106929.2 | NP_001100399.1 | 20, 21 | 2249 | ||
| GenBank mRNA | FM088301.1 | - | 1, 2 | 173 | ||
| FM100327.1 | - | 1, 2 | 173 | |||
| FM120771.1 | - | 4, 5 | 481 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Copg2| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106929.2 | NP_001100399.1 | 20, 21 | 2249 | ||
| GenBank mRNA | FM088301.1 | - | 1, 2 | 173 | ||
| FM100327.1 | - | 1, 2 | 173 | |||
| FM120771.1 | - | 4, 5 | 481 | |||