siRNA Details
| Gene Symbol: RGD1564712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106931.1 | NP_001100401.1 | 2 | 149 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564712| Gene Symbol: RGD1564712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106931.1 | NP_001100401.1 | 2 | 149 | ||