siRNA Details
| Gene Symbol: Zfp711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730265.4 | XP_002730311.1 | 3 | 716 | ||
| XM_008773361.1 | XP_008771583.1 | 5 | 1789 | |||
| XM_008773362.1 | XP_008771584.1 | 4 | 1535 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp711| Gene Symbol: Zfp711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730265.4 | XP_002730311.1 | 3 | 716 | ||
| XM_008773361.1 | XP_008771583.1 | 5 | 1789 | |||
| XM_008773362.1 | XP_008771584.1 | 4 | 1535 | |||