siRNA Details
| Gene Symbol: RGD1563590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257014.2 | XP_006257076.2 | 1 | 243 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1563590| Gene Symbol: RGD1563590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257014.2 | XP_006257076.2 | 1 | 243 | ||