siRNA Details
| Gene Symbol: Mbnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106949.1 | NP_001100419.1 | 2 | 398 | ||
| XM_008773556.1 | XP_008771778.1 | 2 | 1877 | |||
| XM_008773557.1 | XP_008771779.1 | 2 | 1000 | |||
| XM_008773558.1 | XP_008771780.1 | 2 | 997 | |||
| XM_008773559.1 | XP_008771781.1 | 2 | 1000 | |||
| XM_008773560.1 | XP_008771782.1 | 2 | 1003 | |||
| XM_008773561.1 | XP_008771783.1 | 2 | 1000 | |||
| XM_008773562.1 | XP_008771784.1 | 2 | 273 | |||

Mbnl3
Genomic Map