siRNA Details
| Gene Symbol: Rap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106950.2 | NP_001100420.1 | 1 | 252 | ||
| GenBank mRNA | BP503228.1 | - | 1 | 266 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rap2c| Gene Symbol: Rap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106950.2 | NP_001100420.1 | 1 | 252 | ||
| GenBank mRNA | BP503228.1 | - | 1 | 266 | ||