siRNA Details
Gene Symbol: Atp6ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007091.1 | NP_001007092.1 | 9 | 1089 | ||
GenBank mRNA | AB188298.1 | - | 9 | 1089 | ||
BC079339.1 | - | 1089 | 7106562 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atp6ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007091.1 | NP_001007092.1 | 9 | 1089 | ||
GenBank mRNA | AB188298.1 | - | 9 | 1089 | ||
BC079339.1 | - | 1089 | 7106562 |