siRNA Details
| Gene Symbol: Wdr45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013958.1 | NP_001013980.1 | 9 | 777 | ||
| XM_006256710.2 | XP_006256772.1 | 9 | 949 | |||
| XM_006256712.2 | XP_006256774.1 | 9 | 1060 | |||
| XM_006256713.2 | XP_006256775.1 | 9 | 834 | |||
| XM_006256714.2 | XP_006256776.1 | 9 | 954 | |||
| XM_008773071.1 | XP_008771293.1 | 9 | 847 | |||
| GenBank mRNA | BC085816.1 | - | 9 | 777 | 7123612 | |
| FQ225212.1 | - | 4 | 441 | |||

Wdr45
Genomic Map