siRNA Details
Gene Symbol: LOC102557137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773237.1 | XP_008771459.1 | 1 | 1548 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102557137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773237.1 | XP_008771459.1 | 1 | 1548 |