siRNA Details
Gene Symbol: RGD1563378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773242.1 | XP_008771464.1 | 1 | 296 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1563378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773242.1 | XP_008771464.1 | 1 | 296 |