siRNA Details
| Gene Symbol: Rsl1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008876.2 | NP_001008876.1 | 4 | 276 | ||
| GenBank mRNA | AY310144.1 | - | 4 | 276 | ||
| Gene Symbol: NEWGENE_1359295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767502.1 | XP_008765724.1 | 849 | |||

Rsl1d1
Genomic Map