siRNA Details
| Gene Symbol: Ubald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007668.1 | NP_001007669.1 | 3 | 293 | ||
| GenBank mRNA | BC079459.1 | - | 3 | 293 | 7121902 | |
| FQ214502.1 | - | 3 | 330 | |||
| FQ228825.1 | - | 3 | 321 | |||
| Gene Symbol: LOC100910875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750772.3 | XP_003750820.1 | 991 | |||

Ubald1
Genomic Map