siRNA Details
Gene Symbol: Tnfrsf12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181086.3 | NP_851600.1 | 2 | 178 | ||
GenBank mRNA | AY255102.1 | - | 2 | 149 | ||
BC060537.1 | - | 2 | 182 | 6887545 | ||
FQ229590.1 | - | 2 | 178 |
