siRNA Details
| Gene Symbol: Paqr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017377.1 | NP_001017377.1 | 2 | 689 | ||
| XM_006245871.2 | XP_006245933.1 | 2 | 437 | |||
| GenBank mRNA | BC092635.1 | - | 2 | 689 | 7315721 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Paqr4| Gene Symbol: Paqr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017377.1 | NP_001017377.1 | 2 | 689 | ||
| XM_006245871.2 | XP_006245933.1 | 2 | 437 | |||
| GenBank mRNA | BC092635.1 | - | 2 | 689 | 7315721 | |