siRNA Details
| Gene Symbol: Abca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246039.2 | XP_006246101.1 | 24 | 4152 | ||
| XM_220219.9 | XP_220219.6 | 23 | 4011 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abca3| Gene Symbol: Abca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246039.2 | XP_006246101.1 | 24 | 4152 | ||
| XM_220219.9 | XP_220219.6 | 23 | 4011 | |||