siRNA Details
| Gene Symbol: Surf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015014.1 | NP_001015014.1 | 5 | 723 | ||
| XM_006233784.1 | XP_006233846.1 | 5 | 685 | |||
| GenBank mRNA | BC091270.1 | - | 5 | 723 | 7381547 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Surf6| Gene Symbol: Surf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015014.1 | NP_001015014.1 | 5 | 723 | ||
| XM_006233784.1 | XP_006233846.1 | 5 | 685 | |||
| GenBank mRNA | BC091270.1 | - | 5 | 723 | 7381547 | |