siRNA Details
| Gene Symbol: Zfp692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126272.1 | NP_001119744.1 | 3 | 735 | ||
| XM_006246383.2 | XP_006246445.1 | 8 | 1446 | |||
| XM_006246384.2 | XP_006246446.1 | 8 | 1387 | |||
| XM_006246385.2 | XP_006246447.1 | 8 | 1134 | |||
| XM_006246386.2 | XP_006246448.1 | 8 | 1029 | |||
| GenBank mRNA | BC166433.1 | - | 8 | 1020 | 7191618 | |

Zfp692
Genomic Map