siRNA Details
| Gene Symbol: Guk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013115.2 | NP_001013133.2 | 4 | 247 | ||
| XM_006246477.2 | XP_006246539.1 | 5 | 384 | |||
| XM_006246478.2 | XP_006246540.1 | 4 | 349 | |||
| XM_006246479.2 | XP_006246541.1 | 3 | 192 | |||
| XM_006246483.1 | XP_006246545.1 | 5 | 299 | |||
| XM_008767812.1 | XP_008766034.1 | 5 | 433 | |||
| XM_008767813.1 | XP_008766035.1 | 4 | 1008 | |||
| XM_008767814.1 | XP_008766036.1 | 4 | 257 | |||
| XM_008767815.1 | XP_008766037.1 | 4 | 1004 | |||
| GenBank mRNA | AF354443.1 | - | 4 | 198 | ||
| FM065046.1 | - | 4 | 247 | |||

Guk1
Genomic Map