siRNA Details
| Gene Symbol: Slc5a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107007.1 | NP_001100477.1 | 3, 4 | 283 | ||
| XM_008767820.1 | XP_008766042.1 | 3, 4 | 483 | |||
| GenBank mRNA | BC161867.1 | - | 3, 4 | 476 | 7107733 | |

Slc5a10
Genomic Map