siRNA Details
| Gene Symbol: Ulk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191645.1 | NP_001178574.1 | 23 | 3122 | ||
| XM_006246497.2 | XP_006246559.1 | 23 | 3103 | |||
| XM_006246498.2 | XP_006246560.1 | 23 | 3102 | |||
| XM_006246499.2 | XP_006246561.1 | 22 | 2966 | |||

Ulk2
Genomic Map