siRNA Details
| Gene Symbol: Ulk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191645.1 | NP_001178574.1 | 26 | 3558 | ||
| XM_006246498.2 | XP_006246560.1 | 26 | 3538 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ulk2| Gene Symbol: Ulk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191645.1 | NP_001178574.1 | 26 | 3558 | ||
| XM_006246498.2 | XP_006246560.1 | 26 | 3538 | |||