siRNA Details
Gene Symbol: Zfp287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107008.2 | NP_001100478.1 | 4 | 646 | ||
XM_006246692.2 | XP_006246754.1 | 5 | 1066 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107008.2 | NP_001100478.1 | 4 | 646 | ||
XM_006246692.2 | XP_006246754.1 | 5 | 1066 |