siRNA Details
Gene Symbol: Scarf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107022.1 | NP_001100492.1 | 1880 | |||
GenBank mRNA | BC166488.1 | - | 2485 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Scarf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107022.1 | NP_001100492.1 | 1880 | |||
GenBank mRNA | BC166488.1 | - | 2485 |