siRNA Details
| Gene Symbol: Fam57a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727796.4 | XP_002727842.2 | 5 | 2284 | ||
| GenBank mRNA | FQ213392.1 | - | 1 | 18 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam57a| Gene Symbol: Fam57a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727796.4 | XP_002727842.2 | 5 | 2284 | ||
| GenBank mRNA | FQ213392.1 | - | 1 | 18 | ||