siRNA Details
| Gene Symbol: Wsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025664.1 | NP_001020835.2 | 2 | 659 | ||
| NM_001042561.1 | NP_001036026.1 | 7 | 1071 | |||
| XM_008768016.1 | XP_008766238.1 | 7 | 1994 | |||
| GenBank mRNA | BC098720.1 | - | 2 | 659 | ||
| BC167746.1 | - | 7 | 1074 | 8364583 | ||
| CK473503.1 | - | 5 | 554 | |||

Wsb1
Genomic Map