siRNA Details
| Gene Symbol: Ssh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107024.1 | NP_001100494.1 | 13 | 1610 | ||
| XM_006246905.2 | XP_006246967.1 | 14 | 1526 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssh2| Gene Symbol: Ssh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107024.1 | NP_001100494.1 | 13 | 1610 | ||
| XM_006246905.2 | XP_006246967.1 | 14 | 1526 | |||