siRNA Details
| Gene Symbol: Rnf135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012010.1 | NP_001012010.1 | 5 | 895 | ||
| GenBank mRNA | AY539935.1 | - | 854 | |||
| BC087718.1 | - | 5 | 895 | 7316530 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf135| Gene Symbol: Rnf135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012010.1 | NP_001012010.1 | 5 | 895 | ||
| GenBank mRNA | AY539935.1 | - | 854 | |||
| BC087718.1 | - | 5 | 895 | 7316530 | ||