siRNA Details
| Gene Symbol: Hoxb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047091.1 | NP_001040556.1 | 1032 | |||
| GenBank mRNA | DQ399532.1 | - | 1032 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxb2| Gene Symbol: Hoxb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047091.1 | NP_001040556.1 | 1032 | |||
| GenBank mRNA | DQ399532.1 | - | 1032 | |||