siRNA Details
| Gene Symbol: Hoxb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_220896.6 | XP_220896.4 | 2 | 878 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxb1| Gene Symbol: Hoxb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_220896.6 | XP_220896.4 | 2 | 878 | ||