siRNA Details
| Gene Symbol: Smarce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024993.1 | NP_001020164.1 | 10 | 1070 | ||
| XM_008768051.1 | XP_008766273.1 | 10 | 1412 | |||
| GenBank mRNA | BC091314.1 | - | 10 | 1120 | 7312354 | |
| BC092210.1 | - | 10 | 1070 | 7311688 | ||
| FQ213000.1 | - | 10 | 1059 | |||

Smarce1
Genomic Map