siRNA Details
| Gene Symbol: Krt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008752.1 | NP_001008752.1 | 2 | 520 | ||
| XM_006247309.2 | XP_006247371.1 | 2 | 598 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt16| Gene Symbol: Krt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008752.1 | NP_001008752.1 | 2 | 520 | ||
| XM_006247309.2 | XP_006247371.1 | 2 | 598 | |||