siRNA Details
| Gene Symbol: Dhx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098788.1 | NP_001092258.1 | 8 | 1307 | ||
| XM_006247314.2 | XP_006247376.1 | 9 | 1876 | |||
| XM_006247315.2 | XP_006247377.1 | 9 | 1865 | |||
| GenBank mRNA | CB546277.1 | - | 3 | 370 | ||
| FQ228119.1 | - | 2 | 307 | |||

Dhx58
Genomic Map