siRNA Details
Gene Symbol: Arl4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107052.1 | NP_001100522.1 | 2 | 533 | ||
XM_008768053.1 | XP_008766275.1 | 2 | 1519 | |||
XM_008768054.1 | XP_008766276.1 | 3 | 1379 | |||
GenBank mRNA | BC167047.1 | - | 2 | 508 | 8367403 | |
EF688597.1 | - | 2 | 508 |
