siRNA Details
| Gene Symbol: Tmub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031651.2 | NP_001026821.1 | 2 | 625 | ||
| XM_006247331.2 | XP_006247393.1 | 2 | 282 | |||
| XM_006247332.2 | XP_006247394.1 | 3 | 280 | |||
| GenBank mRNA | BC085794.1 | - | 2 | 625 | 7109581 | |
| BC099074.1 | - | 2 | 626 | 7109079 | ||

Tmub2
Genomic Map