siRNA Details
| Gene Symbol: RGD1563746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247481.2 | XP_006247543.1 | 6 | 2609 | ||
| XM_220997.8 | XP_220997.5 | 6 | 2669 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1563746| Gene Symbol: RGD1563746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247481.2 | XP_006247543.1 | 6 | 2609 | ||
| XM_220997.8 | XP_220997.5 | 6 | 2669 | |||