siRNA Details
| Gene Symbol: Kif18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039019.1 | NP_001034108.1 | 3 | 572 | ||
| XM_006247505.2 | XP_006247567.1 | 3 | 580 | |||
| XM_008768283.1 | XP_008766505.1 | 3 | 581 | |||
| GenBank mRNA | BC099126.1 | - | 3 | 572 | 7455959 | |

Kif18b
Genomic Map