siRNA Details
| Gene Symbol: Bptf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC160926.1 | - | 2 | 500 | 8364269 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bptf| Gene Symbol: Bptf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC160926.1 | - | 2 | 500 | 8364269 | |