siRNA Details
| Gene Symbol: Enpp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012466.1 | NP_001012484.1 | 3 | 479 | ||
| GenBank mRNA | AY568760.1 | - | 3 | 479 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Enpp7| Gene Symbol: Enpp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012466.1 | NP_001012484.1 | 3 | 479 | ||
| GenBank mRNA | AY568760.1 | - | 3 | 479 | ||