siRNA Details
Gene Symbol: Igll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190341.1 | NP_001177270.1 | 3 | 466 | ||
XM_006248656.2 | XP_006248718.1 | 3 | 438 | |||
GenBank mRNA | Z68145.1 | - | 3 | 360 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Igll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190341.1 | NP_001177270.1 | 3 | 466 | ||
XM_006248656.2 | XP_006248718.1 | 3 | 438 | |||
GenBank mRNA | Z68145.1 | - | 3 | 360 |