siRNA Details
| Gene Symbol: Dvl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107081.2 | NP_001100551.1 | 12 | 1091 | ||
| XM_006248564.2 | XP_006248626.1 | 12 | 1322 | |||
| XM_006248565.2 | XP_006248627.1 | 12 | 1247 | |||
| XM_006248566.2 | XP_006248628.1 | 12 | 1247 | |||

Dvl3
Genomic Map