siRNA Details
| Gene Symbol: Igf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270598.1 | NP_001257527.1 | 6 | 516 | ||
| NM_001270599.1 | NP_001257528.1 | 6 | 516 | |||
| GenBank mRNA | FQ222567.1 | - | 6 | 284 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igf2bp2| Gene Symbol: Igf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270598.1 | NP_001257527.1 | 6 | 516 | ||
| NM_001270599.1 | NP_001257528.1 | 6 | 516 | |||
| GenBank mRNA | FQ222567.1 | - | 6 | 284 | ||