siRNA Details
| Gene Symbol: Atp13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191657.1 | NP_001178586.1 | 10, 11 | 1228 | ||
| XM_008768800.1 | XP_008767022.1 | 10, 11 | 1253 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp13a5| Gene Symbol: Atp13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191657.1 | NP_001178586.1 | 10, 11 | 1228 | ||
| XM_008768800.1 | XP_008767022.1 | 10, 11 | 1253 | |||