siRNA Details
| Gene Symbol: Osbpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107090.2 | NP_001100560.1 | 10 | 2269 | ||
| XM_006248450.2 | XP_006248512.1 | 9 | 2324 | |||
| XM_006248451.2 | XP_006248513.1 | 10 | 1900 | |||
| XM_006248452.2 | XP_006248514.1 | 10 | 1781 | |||
| XM_008768726.1 | XP_008766948.1 | 10 | 2050 | |||
| GenBank mRNA | BC168952.1 | - | 10 | 2269 | 7453666 | |

Osbpl11
Genomic Map