siRNA Details
| Gene Symbol: Sema5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107091.1 | NP_001100561.1 | 16 | 2564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sema5b| Gene Symbol: Sema5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107091.1 | NP_001100561.1 | 16 | 2564 | ||