siRNA Details
| Gene Symbol: Dirc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012017.1 | NP_001012017.1 | 5, 6 | 1208 | ||
| GenBank mRNA | BC081960.1 | - | 5, 6 | 1208 | 7124589 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dirc2| Gene Symbol: Dirc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012017.1 | NP_001012017.1 | 5, 6 | 1208 | ||
| GenBank mRNA | BC081960.1 | - | 5, 6 | 1208 | 7124589 | |