siRNA Details
| Gene Symbol: Upk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024253.1 | NP_001019424.1 | 5 | 479 | ||
| XM_006248367.2 | XP_006248429.1 | 5 | 686 | |||
| XM_006248368.2 | XP_006248430.1 | 5 | 526 | |||
| GenBank mRNA | BC093613.1 | - | 5 | 479 | 7309649 | |

Upk1b
Genomic Map