siRNA Details
| Gene Symbol: Morc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107109.1 | NP_001100579.1 | 14 | 1713 | ||
| XM_006248060.1 | XP_006248122.1 | 15 | 2046 | |||
| XM_006248061.2 | XP_006248123.1 | 14 | 1972 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Morc3| Gene Symbol: Morc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107109.1 | NP_001100579.1 | 14 | 1713 | ||
| XM_006248060.1 | XP_006248122.1 | 15 | 2046 | |||
| XM_006248061.2 | XP_006248123.1 | 14 | 1972 | |||