siRNA Details
| Gene Symbol: Urb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191661.1 | NP_001178590.1 | 38 | 6515 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Urb1| Gene Symbol: Urb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191661.1 | NP_001178590.1 | 38 | 6515 | ||