siRNA Details
| Gene Symbol: Rnf216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107122.1 | NP_001100592.1 | 14, 15 | 2267 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf216| Gene Symbol: Rnf216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107122.1 | NP_001100592.1 | 14, 15 | 2267 | ||