siRNA Details
| Gene Symbol: Chst12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037775.2 | NP_001032864.1 | 1 | 1223 | ||
| XM_006248935.1 | XP_006248997.1 | 2 | 1317 | |||
| XM_006248936.2 | XP_006248998.1 | 2 | 2366 | |||
| XM_006248937.2 | XP_006248999.1 | 2 | 3104 | |||
| XM_008768971.1 | XP_008767193.1 | 3 | 2248 | |||
| GenBank mRNA | BC100096.1 | - | 1 | 1227 | 7375118 | |

Chst12
Genomic Map